Learn More
DWK Life Sciences DURAN™ Slit sieve
For use with paper or membrane filters.
Brand:  DWK Life Sciences 213405109
Code : NEW
Additional Details : Weight : 0.15500kg
 View more versions of this product
Product Code. 10598991
  • £124.00 / Each
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Description
Description
DURAN™ slotted screen plates serve as support surfaces for the actual filter (e.g. paper or membrane filter), e.g. in the case of buechner funnels.
  • As support for paper or membrane filters
Specifications
Specifications
Borosilicate Glass (3.3)
Suitable for coarse filtration without additional filter medium; Filtration
1 Each
120 mm