Learn More
DWK Life Sciences DURAN™ Slit sieve
For use with paper or membrane filters.
Brand: DWK Life Sciences 213403905
Code : NEW
Additional Details : Weight : 0.52000kg
Description
DURAN™ slotted screen plates serve as support surfaces for the actual filter (e.g. paper or membrane filter), e.g. in the case of buechner funnels.
- As support for paper or membrane filters
Specifications
Borosilicate Glass (3.3) | |
Suitable for coarse filtration without additional filter medium; Filtration | |
10 | |
73 mm |