Learn More
DWK Life Sciences DURAN™ TILT Bottle Tag, GL 56, Silicone Rubber
Simple colour coding for DURAN™ TILT bottles.
Brand:  DWK Life Sciences 292435634
28.76 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.18000kg
 View more versions of this product
Product Code. 15513547
  • £31.70 / Pack of 20
Estimated Shipment: 11-09-2024
to see stock.
Add to basket
Description
Description
The Bottle Tags can be used on their own for colour identification or to secure the protective Light Shield around the bottle. The GL 56 Bottle Tags are available in four colours: orange, yellow, blue and purple.
  • The Bottle Tags can be used for colour identification or to secure the protective Light Shield around the bottle
  • The elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
  • Feature a graspable tab that makes them easy to remove with bare or gloved hands
  • Chemically robust, heat resistant and reusable
Specifications
Specifications
Yellow
color coding and identification; Attachment of the protective Light Shield
GL 56
Silicone
20/Pk.
Yes