Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottle Heads, with Filter Disc
Provides an inexpensive but effective method for the washing and drying of gases. Lenz Laborglasinstrumente™ Gas Washing Bottle Heads, with Filter Disc are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05510401
Code : A0
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11866104
  • £54.06 / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12596, with cone
  • For bottles acc. to DIN 12463
  • Available in a range of porosities
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Duran Tubing
Filter disc
NS 29/32
1 Pack
Gas Washing Bottle Head with Filter Disc
1
DIN 12596, DIN 12463