missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ DURAN™ YOUTILITY™ GL 45 Bottle Tag

Simple color coding for DURAN™ YOUTILITY™ bottles.

Brand:  DWK Life Sciences™ 292432842

35.31 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15795138

  • £42.05 / Pack of 20
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

These colorful GL 45 Bottle Tags offer an elegant solution for the marking of laboratory bottles.

  • Available in different colors for marking of laboratory bottles and personalization
  • Elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
  • Feature a graspable tab that makes them easy to remove with bare or gloved hands
  • Chemically robust, heat resistant and totally reusable
  • Fully compatible with all the GL 45 threads and therefore highly versatile
Specifications

Specifications

20 Tags/Pk.
Green
Color Coding and Identification
Tag
Silicone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers