Learn More
3M™ Dustmaster™ Particulate Filters
Particulate filter for use with the Dustmaster Powered Air Respirator helps protects against dust, mist and fume.
Brand: 3M™ 7000059866
Code : NEW
Additional Details : Weight : 1.03400kg
Description
- Combines with the 6000 Series Gas Vapor cartridges to provide particulate and gas/vapor protection
- Note: 3M 501 filter retainers needed to combine the 5000 and 6000 series
Specifications
P3 | |
6000 and 7000 series facemasks | |
5/Pk. |
Dust Filter | |
Filter |