Learn More
3M™ Dustmaster™ Particulate Filters
Particulate filter for use with the Dustmaster Powered Air Respirator helps protects against dust, mist and fume.
£36.08 - £85.90
Product Code | Brand | Filter Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | Quantity | Price | Quantity & Availability | ||||
15719017
|
3M™
7000037674 |
P2R |
£85.90
10 pairs |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
15719797
|
3M™
7000059865 |
P2 | 5/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12364659
|
3M™
7000006601 |
P2 | 1 Ea. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15729797
|
3M™
7000059866 |
P3 | 5/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Combines with the 6000 Series Gas Vapor cartridges to provide particulate and gas/vapor protection
- Note: 3M 501 filter retainers needed to combine the 5000 and 6000 series