missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Duzmor™ Plus 87-600 Series Natural Rubber Latex Immersion Gloves
A unique combination of strength and elasticity for safer handling of more hazardous substances. Only 0.43 mm thick for outstanding sensitivity and tactile performance.Reduced risk of allergic reactions.
Brand: Ansell™ 87600100BP
20.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Made from unsupported natural rubber
- Reduces risk of allergic reactions
- Unflocked liner
- Fishscale grip
- Bisque fingertip grip
- Pinked cuff

Specifications
Rubber Latex | |
Natural Rubber Latex | |
No | |
10 | |
0.43 mm | |
Pinked | |
12 Pairs | |
87-600 |
Natural Rubber Latex | |
Yes | |
Immersion Gloves | |
87-600 | |
Natural | |
Unflocked | |
Hazardous substances |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell™ Duzmor™ Plus 87-600 Series Natural Rubber Latex Immersion Gloves > Size: 10
Spot an opportunity for improvement?Share a Content Correction