missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Duzmor™ Plus 87-600 Series Natural Rubber Latex Immersion Gloves
A unique combination of strength and elasticity for safer handling of more hazardous substances. Only 0.43 mm thick for outstanding sensitivity and tactile performance.Reduced risk of allergic reactions.
£28.75
Specifications
Back Material | Rubber Latex |
---|---|
Cuff Material | Natural Rubber Latex |
Coating Material | Natural Rubber Latex |
Laboratory/Cleanroom Use | Yes |
Latex Free | No |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
12697021
|
Ansell™
87600070BP |
7 |
£28.75
12 pairs |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
12607031
|
Ansell™
87600080BP |
8 |
£28.75
12 pairs |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
12627031
|
Ansell™
87600100BP |
10 |
£28.75
12 pairs |
In Stock
Log in to see stock. |
||||||
12617031
|
Ansell™
87600090BP |
9 |
£28.75
12 pairs |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
Description
- Made from unsupported natural rubber
- Reduces risk of allergic reactions
- Unflocked liner
- Fishscale grip
- Bisque fingertip grip
- Pinked cuff
Specifications
Rubber Latex | |
Natural Rubber Latex | |
No | |
87-600 | |
Natural | |
Unflocked | |
Hazardous substances |
Natural Rubber Latex | |
Yes | |
Immersion Gloves | |
0.43 mm | |
Pinked | |
12 Pairs | |
87-600 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell™ Duzmor™ Plus 87-600 Series Natural Rubber Latex Immersion Gloves