missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Duzmor™ Plus 87-600 Series Natural Rubber Latex Immersion Gloves

A unique combination of strength and elasticity for safer handling of more hazardous substances. Only 0.43 mm thick for outstanding sensitivity and tactile performance.Reduced risk of allergic reactions.

Brand:  Ansell™ 87600070BP

20.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12697021

  • £28.75 / 12 pairs
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made from unsupported natural rubber
  • Reduces risk of allergic reactions
  • Unflocked liner
  • Fishscale grip
  • Bisque fingertip grip
  • Pinked cuff
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rubber Latex
Natural Rubber Latex
No
7
0.43 mm
Pinked
12 Pairs
87-600
Natural Rubber Latex
Yes
Immersion Gloves
87-600
Natural
Unflocked
Hazardous substances
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ Duzmor™ Plus 87-600 Series Natural Rubber Latex Immersion Gloves > Size: 7

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.