missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ SARS-CoV-2 Spike

The Dynabeads™ SARS-CoV-2 Spike consists of superparamagnetic beads coupled with a trimeric spike protein. The spike protein is a patented recombinant trimeric structure combining the S1 subunit, including the receptor binding domain (RBD), and the S2 subunit found on the surface of the SARS-CoV-2 virus.

Brand:  Invitrogen™ 18100DFIVE

Additional Details : Weight : 0.09350kg

 View more versions of this product

Product Code. 17727443

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The Dynabeads SARS-CoV-2 Spike consists of superparamagnetic beads coupled with a trimeric spike protein. The spike protein is a patented recombinant trimeric structure combining the S1 subunit, including the receptor binding domain (RBD), and the S2 subunit found on the surface of the SARS-CoV-2 virus. The virus spike protein binds to human host cells via the angiotensin-converting enzyme 2 (ACE2) receptor to initiate virus attachment, as well as to antibodies produced in the body as a response to SARS-CoV-2 infection.

Dynabeads SARS-CoV-2 Spike features include:
• Fast SARS-CoV-2 antibody binding kinetics
• Dynabeads-facilitated automation (limited hands-on time)
• Also available in SARS-CoV-2 ELISA kits

This Dynabeads SARS-CoV-2 Spike is also included in our enzyme‐linked immunosorbent assay (ELISA) kits designed to detect and quantify the level of human SARS-CoV-2 IgG (add link to #18000D), IgM (add ink to #18010D), and Total Ig (add link to #18020D) antibodies from serum and plasma samples. These kits can be used manually with a DynaMag-96 magnet or in an automated fashion using a KingFisher instrument. The magnetic beads significantly improve the binding kinetics to cut the ELISA incubation time in half (45 minutes) compared to the more conventional plate-based ELISAs (90 minutes).

Since the spike protein binds host cells to initiate the entrance of SARS-CoV-2 virus into the epithelial cells of the lungs via ACE2, the beads may also be used for isolation of ACE2+ cells or for ACE2 protein purification.

Specifications

Specifications

ISO 13485
The beads are supplied in 0.05 M Tris-buffered saline (TBS) with 0.001% Tween-20 and 0.02% NaN3.

Store at 2°C to 8°C.

Brown
Magnetic Polystyrene
5 x 3 mL
Yes, on KingFisher™ Flex or Apex instruments
Wet ice (2° to 8°C)
2.7 μm
1 mg/mL
Absorbance at 450 nm
Isolation of SARS-CoV-2 Cells
SARS-CoV-2 Spike Protein Magnetic Beads
KingFisher™ Flex or Apex Instruments, DynaMag™-96 Side Skirted, Magnetic 96-Well Separator, ELISA Plate Reader (450 nm)
Dynabeads™ magnetic beads
5 x 3 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only. Not for use in diagnostic procedures.