Learn More
Invitrogen™ Dynabeads™ M-280 Tosylactivated
Description
When you wish to covalently couple antibodies, peptides, intact proteins and functional enzymes to the surface of a magnetic bead choose Dynabeads Tosylactivated. Optimal antibody orientation makes these beads an excellent choice for the immunoprecipitation of proteins and protein complexes.
Ideal for Immunoprecipitation of protein complexes:
Low background and covalent linking of antibodies to the bead surface make Dynabeads Tosylactivated an excellent choice for Immunoprecipitation of proteins and protein complexes (Co-Immunoprecipitation, Co-IP). Gentle and rapid magnetic concentration of the beads and short incubation times (possible due to fast surface-based binding kinetics) make Dynabeads Tosylactivated an excellent choice for the immunoprecipitation of highly-labile or and/or transient (short-lived) protein complexes.
Ultra rapid protocols:
• Identify members of protein complexes in minutes rather than hours
• Temporal resolution short enough to identify transient and labile complexes
• Identify binding partners that cannot be identified with longer protocols
• Rapid protocols further reduce the already ultra-low background binding
• Stronger signal to noise-ratios
Applications:
• IP of proteins and protein complexes
• Couple functional enzymes to bead surface for downstream assays
• Couple peptides to bead surface to identify binders
Purify peptides, proteins and enzymes that are:
• Transiently stable
• Structurally intact
• Temperature labile
• In their native conformation and functional
Coupling Procedure Outline:
Covalent coupling is performed overnight by incubating the desired ligand with the Dynabeads Tosylactivated. Ligands commonly coupled to Dynabeads Tosylactivated include peptides and proteins (e.g. antibodies for Immunoprecipitation or Co-Immunoprecipitation). Coupling occurs at neutral to high pH and at 37°C. We recommend coupling at pH 8.5–9.5, but for pH labile ligands, coupling can be performed in an alternative buffer at pH 7.4.
Upon completion of the ligand coupling step, the actual Dynabeads Tosylactivated surface coating will be rendered inert, resulting in low non-specific binding.
About Dynabeads:
Dynabeads are non-porous monodisperse superparamagnetic beads. They are highly mobile in solution enabling ligands coupled to the beads to continuously interact with the entire sample volume. The superparamagnetic beads are pulled to the tube walls by transferring the tube to a rack containing a strong magnetic field (From the red navigation bar navigate Brands > Dynal > Magnets). Strong magnetic fields quickly pull the beads to the tube wall, allowing for easy and complete removal of the supernatant by pipette. Washing steps are performed similarly.l
Bead Surface Characteristics for Dynabeads Tosylactivated:
• p-toluene-sulfonyl (Tosyl) groups
• Hydrophobic, pH Neutral
• Covalent binding by primary amine (NH2) or sulphydryl (SH) groups
Binding capacity per milligram beads:
Varies depending on ligand, (e.g. 5–10 μg IgG)
Order Info
Shipping Condition: Room Temperature
Warranty
General Terms and Conditions
Specifications
Specifications
| Content And Storage | Store beads at 2°C to 8°C. |
| Elution Volume Min | 5 μL |
| High-throughput Compatibility | High-throughput Compatible |
| Shipping Condition | Room Temperature |
| Sample Type | Any Sample Type |
| Surface Functionality | p-Toluene-Sulfonyl, Tosylactivated |
| Product Line | DYNAL, Dynabeads |
| Quantity | 2 mL |
| Type | Tosylactivated Bead |
| Format | Beads in suspension |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.