missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DynaMag™-2 Magnet

Product Code. 10723874
Click to view available options
Quantity:
1 Each
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10723874

Brand: Invitrogen™ 12321D

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

One DynaMag-2 magnet

The DynaMag-2 magnet combines a strong magnetic attraction for efficient separations in standard microcentrifuge tubes with a flexible and smart ergonomic design.

The DynaMag-2 magnet combines a strong magnetic attraction for efficient separations in standard microcentrifuge tubes with a flexible and smart ergonomic design.

  • Optimal working volume: 10–2000 μL
  • Holds up to 16 standard 1.5–2 mL microcentrifuge tubes in numbered spaces
  • Good control and visibility of sample
  • Top rack can be quickly removed from magnet in base, ready for resuspension, vortexing, rotation, or manual sample shaking
  • Magnet can be used with all types of Dynabeads for efficient isolation of proteins and nucleic acids
  • Sample rack can be easily removed from base magnet for resuspension, vortexing, rotation or manual sample shaking
  • Center pin in rack ensures equal vortexing of all tubes
  • Rack can also be removed and used to store tubes
  • 16 numbered sample spaces make magnet for parallel handling of multiple samples
  • With eight tube positions on each side of rack, easily scale protocols to accommodate 96-well plate reactions

This magnet is not compatible with the MX4 Mixer. The removable sample rack is also available as a separate item. Purchasing an additional sample rack minimizes the need to move tubes between racks. The rack can be placed on the bench, in the refrigerator, or in the incubator.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Description Magnet
Content And Storage Contains one DynaMag™-2 magnet. Store at room temperature.
Shipping Condition Room Temperature
Product Line DYNAL, DynaMag, Dynabeads
Quantity 1 Each

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.