Learn More
Molecular Probes™ DynaLight™ Trigger Solution
Achieve faster ELISA results with DynaLight™ Trigger Solution when used in combination with DynaLight™ Substrate with RapidGlow™ Enhancer, a ready-to-use chemiluminescent substrate formulation that has been optimized to achieve faster results in solution-based assays.
£116.00 - £504.00
Specifications
Product Type | Trigger Solution |
---|---|
Detection Method | Chemiluminescent |
Form | Liquid |
For Use With (Equipment) | Luminometer (Tube-Based) |
High-throughput Compatibility | High-throughput Compatible |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12807452
|
Molecular Probes™
4475409 |
1 Bottle |
£504.00
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
12887442
|
Molecular Probes™
4475403 |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Features of DynaLight™ Substrate with RapidGlow™ Enhancer:
• Faster kinetics and higher signal levels than colorimetric, fluorescent, or other chemiluminescent chemistries
• Sensitive attomole detection of purified alkaline phosphatase enzyme
• Optimized formulation for both microplate and bead-based assays
• Substrate stability of over 1 year at 4°C
• Compatible with automated platforms with trigger solution/positions already in place
The DynaLight™ Substrate with RapidGlow™ Enhancer provides a robust chemiluminescent reaction when catalyzed by alkaline phosphatase. This reaction achieves maximum light emission within 2 minutes and maintains a sustained signal for hours. When combined with DynaLight™ Trigger Solution, DynaLight™ Substrate with RapidGlow™ Enhancer delivers maximum signal within seconds. The environmentally-friendly DynaLight™ Trigger Solution provides maximum signal within 1 second of addition, with a longer half-life to its ground state than other chemiluminescent Flash technologies, allowing ample time to collect data reliably.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
Trigger Solution | |
Liquid | |
High-throughput Compatible | |
Cell Cultures, Whole Blood, Serum | |
Alkaline Phosphatase | |
Room Temperature |
Chemiluminescent | |
Luminometer (Tube-Based) | |
DynaLight™ | |
Chemical Substrate | |
31 sec. |