Learn More
Invitrogen™ DynaMag™-96 Bottom Magnet
Magnet pulls beads to the bottom of each well. Compatible with PCR-strips and 96-well plates (200 μL) non-skirted and half-skirted and replaces the Dynal MPC-96B.
Brand: Invitrogen™ 12332D
Code : 50
Additional Details : Weight : 0.49000kg
Includes: One magnet
Description
Magnet pulls beads to the bottom of each well. Compatible with PCR-strips and 96-well plates (200 μL) non-skirted and half-skirted and replaces the Dynal MPC-96B. When used in combination with the DynaMag-96 Side on an automation platform, the two magnets allow for significant volume changes through a workflow.
- High wash volumes—DynaMag-96 Side
- Low Elution Volumes—DynaMag-96 Bottom
- Concentrates Beads at the bottom of each well
- Features 96 round magnets (one centered at the bottom of each well: to ensure identical sample handling at each position)
- Allows for concentrating the sample by eluting in tiny volumes (approx. 10μL)
Order Info
Shipping Condition: Room Temperature
Specifications
Magnet | |
DynaMag™ | |
DynaMag™ |
Contains one magnet. Store at room temperature. | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.