missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DynaMag™-96 Side Magnet

Product Code. 10531948
Click to view available options
Quantity:
1 Each
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10531948

Brand: Invitrogen™ 12331D

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

One magnet

Designed for both automation platforms (e.g., Tecan) and manual workflows, but is compatible with PCR-strips and 96-well plates (200 μL), non-skirted and half-skirted.

Designed for both automation platforms (e.g., Tecan) and manual workflows, but is compatible with PCR-strips and 96-well plates (200 μL), non-skirted and half-skirted.

  • High wash volumes: DynaMag-96 Side Magnet
  • Low elution volumes: DynaMag-96 Bottom magnet
  • With extra column (13 total)
  • Enables sample mixing simply by shifting 96-well plate back and forth from right-most position to left-most position
  • Shifting draws beads back and forth through sample and allows mixing to be achieved without handling, promoting faster processing and less sample handing-induced variation
  • When used in combination with DynaMag-96 Bottom magnet on automation platform, two magnets allow for significant volume changes through workflow

Magnet is not compatible with PCR tubes.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Description Magnet
Content And Storage Contains one magnet. Store at room temperature.
For Use With (Equipment) DynaMag™
Shipping Condition Room Temperature
Product Line DynaMag
Quantity 1 Each

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.