missing translation for 'onlineSavingsMsg'
Learn More
Dysprosium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11345929

Dysprosium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11345929
50 x 50 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy

Product Code. 11345929

Brand: Thermo Scientific Alfa Aesar 012382.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 0.53g/25 x 25mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]

Specifications

Color Gray to Silver
Physical Form Foil
Quantity 50 x 50 mm
Assay Percent Range 99.90%
Odor Odorless
Appearance Gray to silver foil
Chemical Name or Material Dysprosium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW24H007 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012382.FF,012382.FI
Lot NumberJ20R048 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012382.FF,012382.FI
Lot NumberL05X027 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012382.FF,012382.FI
Lot NumberE19T012 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012382.FF,012382.FI
Lot NumberY18B011 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012382.FF,012382.FI

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dysprosium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.