Learn More
Cytiva EAH Sepharose™ 4B Medium
Used for coupling compounds containing carboxyl groups to Sepharose 4B. EAH Sepharose™ 4B Medium provides extremely stable coupling via an 11-atom spacer arm.
Brand: Cytiva 17-0569-01
Code : GE
Additional Details : Weight : 0.99790kg
Description
- Group to be coupled: -COOH, -CHO
- Matrix : 4% Agarose
- Average particle size: 90μm
- Extremely stable coupling via an 11-atom hydrophilic spacer arm
Specifications
4% Agarose | |
3 to 14 | |
4°C to 8°C |
90 μm | |
50 mL | |
Used for coupling compounds containing carboxyl groups to Sepharose 4B through Carbodiimide based coupling via an 11-atom spacer arm |
Safety and Handling
- EAH Sepharose 4B, 50 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol