Learn More
Abeba™ Easy 37621 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand: Abeba™ 37621/35
Additional Details : Weight : 0.97000kg
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Microfiber
- Patent Black
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3573)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Microfiber | |
Slip-resistant | |
37621 |
Patent Black | |
35 | |
Polyurethane |