Learn More
Abeba™ Easy 37621 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 37621/39
Additional Details : Weight : 0.97000kg
 View more versions of this product
Product Code. 15681088
  • £60.94 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber
  • Patent Black
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
37621
Patent Black
39
Polyurethane