Learn More
ABEBA™ Easy White Occupationnal Clog OB
ABEBA™ Easy White Occupationnal Clog OB with a fixed heel strap, replaceable ACC Wave insole, and slip resistant TPU outsole.
Brand:  ABEBA™ 37620/42
Additional Details : Weight : 1.05000kg
 View more versions of this product
Product Code. 16388201
  • £60.94 / 1 pair
Estimated Shipment: 06-08-2024
to see stock.
Add to basket
Description
Description
  • Quality, comfort, fitting
  • Comply to EN ISO 20347 : 2012, OB SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • White water and chemical resistant microfiber
  • Foldable heel strap, adjustable
  • Sanitized treatment
Specifications
Specifications
Stainless steel
Heel strap
Microfiber
42
Thermoplastic Polyurethane
1 Pair
Water
CE EN ISO 20347 :2012, OB, A, E, FO, SRC, ESD
White
1
Slip and abrasion resistant, ESD dissipative, shock absorbent
Clogs
EN ISO 20347 : 2012; DIN EN 61340