missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EasyPep™ Magnetic MS Sample Preparation Kits and Proteomics Magnetic Clean-up Beads

EasyPep Magnetic MS Sample Preparation Kits and Proteomics Magnetic Clean-up Beads contain MS-compatible magnetic beads specifically designed to tolerate harsh MS sample prep conditions to improve sample quality and reproducibility for both manual and automated sample preparation methods.

£292.00 - £1358.00

Specifications

Starting Material Protein Samples
Detection Method Mass Spectrometry
Form Liquid
View More Specs

Products 3
Product Code Brand Quantity Product Type Price Quantity & Availability  
Product Code Brand Quantity Product Type Price Quantity & Availability  
18496448
View Documents
Thermo Scientific™
A57866
20 Reactions Magnetic MS Sample Prep Kit
£498.00
Each
Please to purchase this item. Need a web account? Register with us today!
18426548
View Documents
Thermo Scientific™
A57867
96 Reactions Magnetic MS Sample Prep Kit
£1358.00
Each
Please to purchase this item. Need a web account? Register with us today!
18416458
View Documents
Thermo Scientific™
A57868
5 mL Magnetic Clean-up Beads
£292.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Proteomic sample preparation for MS analysis is complex, resulting in challenges with reproducibility and peptide quality. For MS sample preparation methods that utilize magnetic beads, a major challenge is the compatibility of the beads with the harsh conditions required for sample processing. When exposed to extreme pH or solvents, traditional magnetic beads often clump or dissolve, resulting in increased variability.

Pierce Proteomics Magnetic Clean-up Beads have been designed for compatibility with common solvents (e.g. acetonitrile, ethanol, isopropyl alcohol, and acetone) and wide range of pH (e.g. pH 2-13) to enable fast, efficient, and reproducible removal of a wide variety of detergents, salts, and other mass spectrometry (MS)-incompatible contaminants from protein and peptide samples. The beads can be successfully used to clean up proteins derived from different sample types (e.g. purified proteins, cell lysates, tissue extracts, and biological fluids) and protein input amounts (e.g. 1 µg to 1 mg) using manual magnetic processing or automated workflows. 

EasyPep Magnetic MS Sample Prep Kits integrate optimized EasyPep sample preparation chemistry with the Proteomics Magnetic Clean-up Beads to support manual and automated magnetic bead sample processing, including the King Fisher Instrument Series. The complete kits contain pre-formulated buffers, MS-grade enzyme mix, magnetic beads for peptide clean-up, and an optimized protocol to generate MS-ready peptide samples in less than 3 hours. The kits are suitable for processing protein samples from 15–100 µg with a high yield of detergent-free peptides ready for direct LC-MS analysis without the need to SpeedVac dry and reconstitute before injection.  In addition, the peptide samples are compatible with isobaric tag (e.g., TMT/TMTpro Reagents) labeling and high pH reversed-phase fractionation.

Specifications

Specifications

Protein Samples
Mass Spectrometry
Liquid
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ EasyPep™ Magnetic MS Sample Preparation Kits and Proteomics Magnetic Clean-up Beads

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.