Learn More
STEMCELL Technologies EasySep™ Human Monocyte Isolation Kit
Immunomagnetic isolation of untouched human monocytes
Brand:  STEMCELL Technologies 19359
Encompass
Product Code. 16698092
  • £1069.50 / Each
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Human Monocyte Isolation Kit is designed to isolate CD14+CD16- monocytes from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection
  • The EasySep™ procedure involves labeling unwanted cells and platelets with antibody complexes and magnetic particles
  • The magnetically labelled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
  • This product can be used in place of the EasySep™ Human Monocyte Enrichment Kit (Catalog #19059) for even faster cell isolations with reduced platelet contamination
Specifications
Specifications
Monocytes
Cell Isolation
1 x 109 cells