Learn More
STEMCELL Technologies EasySep™ Mouse CD4+CD62L+ T Cell Isolation Kit
Immunomagnetic isolation of mouse naïve CD4+ T cells
Brand:  STEMCELL Technologies 18765
Encompass
Product Code. 17197861
  • £904.50 / 1 set
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Isolate highly purified naïve CD4+ T cells from single-cell suspensions of splenocytes or other tissues by using a simple, two-step method
  • First, naive CD4+ T cells are pre-enriched by negative selection using the EasySep™ Mouse Naïve CD4+ T Cell Pre-Enrichment Cocktail containing biotinylated antibodies directed against non-naïve CD4+ T cells (CD8, CD11b, CD11c, CD19, CD24, CD25, CD45R, CD49b, TCRγ/δ, TER119)
  • Then CD4+CD62L+ cells, pre-labeled with CD62L PE, are selected using the EasySep™ PE Selection Cocktail
  • The desired cells are labeled with antibodies and magnetic particles
  • The cells are separated without columns using an EasySep™ magnet
  • For the isolation of mouse naïve CD4+ T cells in as little as 15 minutes by negative selection, we recommend using the EasySep™ Mouse Naïve CD4+ T Cell Isolation Kit (19765)
Specifications
Specifications
T Cells, T Cells, CD4+
Cell Isolation
1 x 109 cells