Learn More
STEMCELL Technologies EasySep™ Mouse CD8+ T Cell Isolation Kit
Immunomagnetic isolation of untouched mouse CD8+ T cells
Brand:  STEMCELL Technologies 19853
Encompass
Product Code. 17157971
  • £778.50 / 1 set
Estimated Shipment: 11-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Mouse CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection
  • Unwanted cells are targeted for removal with biotinylated antibodies directed against non-CD8+ T cells and streptavidin-coated magnetic particles (RapidSpheres™ )
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
  • This product replaces the EasySep™ Mouse CD8+ T Cell Enrichment Kit (Catalog #19753) for even faster cell isolations
Specifications
Specifications
T Cells, T Cells, CD8+
Cell Isolation
1 x 109 cells