missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Essential Human Th1/Th17 Phenotyping Kit

Invitrogen™ eBioscience™ Essential Human Th1/Th17 Phenotyping Kit is designed for flow cytometry-based identity testing of Type 1 and Type 17 T-helper cells

Brand:  Invitrogen™ A42927

Product Code. 17164729

  • £1034.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Phenotyping Kit leverages high quality eBioscience™ antibodies, isotype controls, cell stimulation reagents (induces cytokine production), and fixation/permeabilization reagents (for intracellular markers IfN-g and IL-17) and includes a ready-to-use protocol for experimental setup, gating strategy, and analysis.

Benefits include:

  • Proven performance―antibodies and supporting reagents designed for flow cytometry
  • Highly referenced antibodies―includes one of the most referenced clones for IL-17
  • Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization
TRUSTED_SUSTAINABILITY
Specifications

Specifications

  • CD4, Ifn-g, IL-17a antibodies, store at 2°C to 8°CC
  • Cell stimulating cocktail, store at -5 to -30°C
  • Protein transport cocktail, store at -5 to -30°C
  • Intracellular fixation & permealization reagents, store at 2°C to 8°CC
  • Isotype control, store at 2°C to 8°CC
Kit
Approved for shipment at Room Temperature or on Wet or Dry Ice
Phenotyping Kit
Fluorescence
100 reactions
eBioscience™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.