missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Essential Human Th1/Th17 Phenotyping Panel with Staining & Stimulation Reagents
Click to view available options
Quantity:
100 reactions
Unit Size:
100 reactions
Description
Phenotyping Panel with Staining and Stimulation Reagents leverages high quality eBioscience™ antibodies, cell stimulation reagents (induces cytokine production), and fixation/permeabilization reagents (for intracellular markers IfN-g and IL-17) and includes a ready-to-use protocol for experimental setup, gating strategy, and analysis.
Benefits include:
- Proven performance―antibodies and supporting reagents designed for flow cytometry
- Highly referenced antibodies―includes one of the most referenced clones for IL-17
- Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization
Specifications
Specifications
| Content And Storage |
|
| Format | Kit |
| Quantity | 100 reactions |
| Detection Method | Fluorescence |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
| Product Line | eBioscience |
| Product Type | Phenotyping Panel |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction