Learn More
Invitrogen™ eBioscience™ Human Cell Surface Flow Cytometry Workflow Kit
The eBioscience™ Human Cell Surface Flow Cytometry Workflow Kit contains all the buffers, blockers, and beads needed to complete a standard flow cytometry experiment for staining cell surface targets on human cell suspensions.
Brand:  Invitrogen™ A53011
Product Code. 17777463
  • £325.00 / Each
Explore more special offers
Description
Description
  • Kit contains all the buffers, blockers, and beads needed to complete a standard flow cytometry experiment for staining cell surface targets on human cell suspensions
  • When combined with experiment-specific antibodies, the reagents within this kit have been optimized to work together to detect cell surface targets by flow cytometry
  • Effective compensation controls
  • Fc receptor blocking
  • Non-antibody, non-protein-based blocking solution for reducing background staining from fluorophores
  • Antibody binding buffer minimizes non-specific binding of antibodies
  • Fixation buffer for storage of stained samples
Kit Includes:
  • UltraComp eBeads™ Plus Compensation Beads (01-3333-41)
  • Fc Receptor Binding Inhibitor Polyclonal Antibody, eBioscience (14-9161-71)
  • CellBlox Monocyte and Macrophage Blocking Buffer (B001T02F01)
  • eBioscience Flow Cytometry Staining Buffer (00-4222-57)
  • eBioscience IC Fixation Buffer (00-8222-49)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Flow Cytometry
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ eBioscience™ Human Cell Surface Flow Cytometry Workflow Kit > 1 Kit