missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ ECO AluRack for Upright Freezers, 50 mm Box Height
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
£73.05 - £99.00
Specifications
Description | Seawater resistant |
---|---|
Holds | Boxes |
Material | Aluminum |
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 50 mm (Box) |
Product Code | Brand | Holds | Array | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Array | Price | Quantity & Availability | |||||
12402510
|
Tenak™
TE11224 |
Boxes | 5 x 3 |
£77.65
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
|||||
12412520
|
Tenak™
TE11238 |
Boxes | 7 x 4 |
£99.00
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
12422510
|
Tenak™
TE11230 |
Boxes | 3 x 4 |
£75.10
Each |
In Stock
Log in to see stock. ![]() |
|||||
12462500
|
Tenak™
TE11220 |
Boxes | 3 x 3 |
£73.05
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
|||||
12482500
|
Tenak™
TE11222 |
Boxes | 4 x 3 |
£75.10
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
|||||
12482520
|
Tenak™
TE11256 |
Boxes | 6 x 5 |
£97.60
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
|||||
15230464
|
Tenak™
TE11250 |
Boxes | 3 x 5 |
£85.85
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
|||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Without white cardboard boxes
- Holds is expressed by Height x Dept
Specifications
Seawater resistant | |
Aluminum | |
50 mm (Box) |
Boxes | |
Upright Freezers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tenak™ ECO AluRack for Upright Freezers, 50 mm Box Height