missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ ECO AluRack for Upright Freezers, 50 mm Box Height
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE11224
This item is not returnable.
View return policy
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Without white cardboard boxes
- Holds is expressed by Height x Dept
Specifications
Seawater resistant | |
Boxes | |
15 | |
425 x 139 x 276 mm |
5 x 3 | |
Aluminum | |
Upright Freezers | |
50 mm (Box) |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tenak™ ECO AluRack for Upright Freezers, 50 mm Box Height > Holds: 15 Boxes (5 x 3)
Spot an opportunity for improvement?Share a Content Correction