Learn More
Seward™ Stomacher™ Bag Racks
Seward has developed a full range of Stomacher™ Accessories to help provide a safe and efficient working environment in your laboratory.
Brand:  Seward™ BA6096
 View more versions of this product
Product Code. 10748898
  • £205.00 / Each
Estimated Shipment: 06-05-2025
Explore more special offers
Add to basket
Description
Description
Help to organize sample bags
  • High-grade plastic polymer construction
  • Provide perfect storage for Stomacher bag sample handling for a cleaner, more productive work environment
  • Perfect for loading closure bags pre-enrichment into the incubator
  • Easily dismantled and cleaned with most lab disinfectants (not autoclavable)
Specifications
Specifications
Bag Rack
Clear
Polymer
Rectangular
To prepare, store and transport samples in microbiology sample preparation
1
35cm x 10cm x 22cm
Not Autoclavable
Bags
10
Non-sterile
Stomacher 400
1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Seward™ Stomacher™ Bag Racks > For Seward Stomacher 400 lab blender; Holds 10 bags