missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ ECO AluRack for Upright Freezers, 50 mm Box Height, with Boxes

ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE11222B

 View more versions of this product

Product Code. 12492500

  • £111.00 / Each
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of seawater resistant aluminum
  • Fits into any freezer and can be used with all standard cryoboxes
  • Racks can be used with all freezer brands
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
  • Holds is expressed by Height x Dept
Specifications

Specifications

White cardboard boxes, 9 x 9 cell dividers
4 x 3
Aluminum
Upright Freezers
50 mm (Box)
Seawater resistant
Boxes
12
425 x 139 x 221 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tenak™ ECO AluRack for Upright Freezers, 50 mm Box Height, with Boxes > Holds: 12 Boxes (4 x 3)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.