Learn More
Sharpsguard™ Eco Anatomical Waste Container
The new Sharpsguard™ eco anatomical container, made using at least 40% recycled plastic with red solid lid is for the disposal of anatomical waste and other potentially infectious waste.
Brand: Sharpsguard™ DD910R
This item is not returnable.
View return policy
Description
- Sharpsguard™ eco anatomical is for the safe disposal of anatomical and other potentially infectious waste.
Specifications
10.91 L | |
274 mm | |
248 mm | |
Safe Disposal of Anatomical and Potentially Infectious Waste |
Red | |
Yellow | |
20/Pk. |