Learn More
LTE Scientific™ Eco-Drying Cabinets
Reduce typical energy usage by up to 50% when compared to traditional drying cabinets. LTE Scientific™ Eco-Drying Cabinets are ideal for drying glassware, instruments or media and can also be used for warming.
Brand:  LTE Scientific™ DC/LOW/100
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15656228
  • £2167.20 / 1 unit
Estimated Shipment: 18-09-2024
to see stock.
Add to basket
Description
Description
  • Insulation is provided to 5 sides of the cabinets
  • Temperature is electronically controlled
  • Temperature is digitally displayed
Specifications
Specifications
100 L
85°C
25 kg
425 x 778 x 302 mm