Learn More
LTE Scientific™ Eco-Drying Cabinets
Reduce typical energy usage by up to 50% when compared to traditional drying cabinets. LTE Scientific™ Eco-Drying Cabinets are ideal for drying glassware, instruments or media and can also be used for warming.
£2167.20 - £2485.00
Specifications
Temperature (Metric) 85°C
View More Specs
Products 2
Product Code Brand Capacity (Metric) Weight (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Weight (Metric) Price Quantity & Availability  
15636228
View Documents Promotion Details
LTE Scientific™
DC/LOW/180
180 L 38 kg
£2485.00
1 unit
View alternative products
Save Up to 
»
Estimated Shipment: 18-09-2024
to see stock.
Add to basket
15656228
View Documents Promotion Details
LTE Scientific™
DC/LOW/100
100 L 25 kg
£2167.20
1 unit
View alternative products
Save Up to 
»
Estimated Shipment: 18-09-2024
to see stock.
Add to basket
Description
Description
  • Insulation is provided to 5 sides of the cabinets
  • Temperature is electronically controlled
  • Temperature is digitally displayed
Specifications
Specifications
85°C