Learn More
Sharpsguard™ Eco EXTRA 8.5 Waste Container
The new Sharpsguard™ eco EXTRA 8.5 container, made using at least 40% recycled plastic that is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic.
Brand: Sharpsguard™ DD580OLR
Additional Details : Weight : 12.24000kg
Description
- Sharpsguard™ eco EXTRA 8.5 is an ideal solution for the management of sharps in high public access areas.
- This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
- This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-01-09, 18-02-01, 18-02-02, 18-02-08.
Especificaciones
7.59 L | |
360 x 161 x 269 mm | |
816 g |
Orange | |
15/Pk. |