Learn More
Sharpsguard™ Eco EXTRA 8.5 Waste Container
GREENER_CHOICE
The new Sharpsguard™ eco EXTRA 8.5 container, made using at least 40% recycled plastic that is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic.
Brand:  Sharpsguard™ DD580YLR
 View more versions of this product
Product Code. 17754864
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Sharpsguard™ eco EXTRA 8.5 is an ideal solution for the management of sharps in high public access areas.
  • This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-01-09, 18-02-01, 18-02-02, 18-02-08.
Specifications
Specifications
7.59 L
360 x 161 x 269 mm
816 g
Yellow
15/Pk.