Learn More
Ecolite(+) LC Fluid, MP Biomedicals
Liquid Scintillation Cocktails; Ecolite(+); MP Biomedical; Biodegradable
Brand:  MP Biomedicals 0188247504
 View more versions of this product
Product Code. 15752252
  • £756.00 / 15.14 L
Explore more special offers
Specifications
Liquid Scintillation Cocktails
4 x 1 gal.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ecolite(+) LC Fluid, MP Biomedicals > 4 x 1 gal.