Learn More
Ansell™ Econohands™ 87-190 Series Yellow Natural Rubber Latex Extra Thin Gloves
Econohands™ 87-190 Series Yellow Natural Rubber Latex Extra Thin Gloves - Extra thin glove for excellent sensitivity. Specially treated to reduce the risk of allergic reactions.
£15.60
Specifications
Back Material Rubber Latex
Cuff Material Natural Rubber Latex
Coating Material Natural Rubber Latex
Latex Free No
Product Type Extra Thin Gloves
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12697011
View Documents
Ansell™
87190100BP
10
£15.60
12 pairs
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
12667011
View Documents
Ansell™
87190070BP
7
£15.60
12 pairs
In Stock
to see stock.
Add to basket
12677011
View Documents
Ansell™
87190080
8
£15.60
12 pairs
In Stock
to see stock.
Add to basket
12687011
View Documents
Ansell™
87190090BP
9
£15.60
12 pairs
In Stock
to see stock.
Add to basket
Description
Description
  • Specially treated to reduce the risk of allergic reactions.
  • Thickness of 0.35 mm, provides outstanding sensitivity.
  • Excellent price/quality ratio.
Specifications
Specifications
Rubber Latex
Natural Rubber Latex
Extra Thin Gloves
0.35 mm
Straight
12 Pairs
87-190
Natural Rubber Latex
No
87-190
Yellow
Cotton Flocking
Liquid & mechanical resistant
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Ansell™ Econohands™ 87-190 Series Yellow Natural Rubber Latex Extra Thin Gloves