Learn More
Fisherbrand™ Mechanical Benchtop Counter
Depress the keys in the appropriate blood cell category to keep track of counts. Mechanical Benchtop Counter is an economical solution for counting blood cells, bacterial colonies and other types of repetitive counting.
Brand:  Fisherbrand™ HS5360B
 View more versions of this product
Product Code. 11344095
  • £327.00 / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Each key records up to 999
  • Total window keeps track of total number of strokes made on all other keys
  • Bell rings when total reaches 100 blood cell key labels
    • HS5360A:
      • Baso., Eosin., Mono., Lymph., Poly., and Total,
      • Measures 250 x 76 x 56 mm
    • HS5360B:
      • Baso, Eosin., Myelo., Juven., Stab., Seg., Lymph., Mono., and Total,
      • Measures 320 x 76 x 56 mm
Specifications
Specifications
Mechanical Counter
9-key Counter
1 Ea.
56 mm
320 mm
76 mm
Mechanical
HS5360B
2.2 in.
12.6 in.
3 in.
Labels: Baso, Eosin., Myelo., Juven., Stab., Seg., Lymph., Mono., and Total