missing translation for 'onlineSavingsMsg'
Learn More

Norell™ Economy Standard Borosilicate Glass NMR Sample Tubes

For auto-sampling systems and high-throughput NMR

Brand:  Norell™ 1001-8

 View more versions of this product

Product Code. 12830026

  • £1130.00 / Pack of 100
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For auto-sampling systems and high-throughput NMR
  • The Norell Standard Series™ of NMR tubes has been developed for quality and precision
  • Virtually all bottom and wall variation has been eliminated, greatly improving shimmering times and sample reproducibility
  • Whether monitoring a reaction, scouting for a new compound, or doing routine NMR, the Standard Series™ is the tube of choice for today's scientist
  • All tubes have polished ends making them less susceptible to breaking
  • Colored caps are included
Specifications

Specifications

203 mm
NMR Sample Tube
10 mm
For auto sampling systems and high-throughput NMR
Borosilicate Glass
100 Pack
8.76 mm
Caps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers