Learn More
Tango™ EDG7-bla U2OS Cells
Tango™ EDG7-bla U2OS Cells contain the human Endothelial Differentiation Gene 7.
Brand: Tango™ K1849
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Accession Number: NM_012152.1
- Cytoplasmic Tail: Native Tail
- Population Status: Single Cell Clone Origin
- Tango™ EDG7-bla U2OS cells contain the human Endothelial Differentiation Gene 7 (EDG7) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ EDG7-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of LPA. In addition, Tango™ EDG7-bla U2OS cells have been tested for assay performance under variable conditions.
Specifications
Liquid Nitrogen | |
GPCRs | |
Dividing Cells |
Fluorescent | |
8 x 106 Cells |