Learn More
Tango™ EDG8-bla U2OS Cells
Tango™ EDG8-bla U2OS Cells contain the human Endothelial Differentiation Gene 8 (EDG8) linked to a TEV protease site.
Brand: Tango™ K1518
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Gene Symbol: EDG8
- Accession Number: NM_030760.3
- Cytoplasmic Tail: Tail from AVPR2 GPCR
- Population Status: Pool of Stable Cells
- The Tango™ EDG8-bla U2OS cells contain the human Endothelial Differentiation Gene 8 (EDG8) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
- The Tango™ EDG8-bla U2OS cells are functionally validated for Z' and EC50 concentrations of S1P. In addition, Tango™ EDG8-bla U2OS cells have been tested for assay performance under variable conditions.
Specifications
Should be stored in liquid nitrogen immediately upon receipt | |
GPCRs | |
8 x 106 Cells |
Fluorescent | |
EDG8-bla U2OS cells, EDG8-bla U2OS Assay Protocol, Certificate of Analysis | |
Dividing Cells |