Learn More
Epredia™ Edge-Rite™ Disposable Microtome Blades
Experience consistent quality from blade to blade and sample to sample with Epredia™ Edge-Rite™ disposable microtome blades.
Brand:  Epredia™ 4275H
155.76 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 12688066
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Surgical-grade steel
  • High- and low-profile blades are specially coated to increase longevity and optimize section quality
Low profile blades:
  • Blades are 20% thicker than traditional blades to increase knife edge stability, which allows for excellent ribboning and facilitates usage in cryostats
  • Blades are 5% longer, allowing for more sections per blade
  • Blades fit any low-profile blade holder
Specifications
Specifications
Disposable Microtome Blade
High
Surgical-grade Steel
Single-use