missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PL BioScience ELAREM™ Perform, Human Platelet Lysate, Heparin Required
ELAREM™ Perform is a human Platelet Lysate of EU origin supporting in vitro expansion of various primary cells and cell lines. The safe and performance-increasing cell growth promoter ensures efficient lab processes with batch-to-batch consistency and a xeno-free cell environment.
Brand: PL BioScience PE10811
This item is not returnable.
View return policy
Description
- Xeno-Free: The xeno-free cell culture supplement is obtained from transfusion-approved platelets obtained at licensed blood centres.
- EU Origin: ELAREM™ Perform Research Grade is manufactured from platelet units obtained from healthy blood donors at licensed blood centres in the EU.
- Improved Cell Performance: Human Platelet Lysate contains abundant growth factors and cytokines derived from human platelets. Those nutrients have been shown to stimulate cellular proliferation and maintain phenotype and differentiation potential of various cells.
- Reproducible Results: Each batch of ELAREM™ Perform is produced from large pools of transfusion-approved platelet units to ensure batch-to-batch consistency and enable reproducible conditions. Grade & GMP GradeB
- From Research to Clinical Development: ELAREM™ Perform meets the requirements of academic and industrial cell expansion and assures an easy transition of research results into clinical applications.
- The product contains coagulation factors. To inhibit coagulation of the complete cell culture medium, it is recommended to add an anticoagulant, e.g. heparin, to the cell culture medium.
- Intended Use: ELAREM™ Perform is for in vitro and manufacturing use only. The product is not intended for direct therapeutic use. For safety precautions, please see appropriate Safety Data Sheet (SDS).
Specifications
Clear, yellow | |
Liquid | |
Sterile | |
Liquid, Human Platelet Lysate | |
100 mL |
Adherent Cell Culture, Suspension Cell Culture | |
Cell Culture Supplement | |
Sterile-filtered | |
Research Grade | |
Dry Ice |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction