missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PL BioScience ELAREM™ Perform, Human Platelet Lysate, Heparin Required
ELAREM™ Perform is a human Platelet Lysate of EU origin supporting in vitro expansion of various primary cells and cell lines. The safe and performance-increasing cell growth promoter ensures efficient lab processes with batch-to-batch consistency and a xeno-free cell environment.
£151.55 - £1558.80
Specifications
Culture Type | Adherent Cell Culture, Suspension Cell Culture |
---|---|
Form | Liquid |
Product Type | Cell Culture Supplement |
Sterility | Sterile |
Sterilization Method | Sterile-filtered |
Product Code | Brand | Quantity | Purity or Quality Grade | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Purity or Quality Grade | Price | Quantity & Availability | ||||
17977731
|
PL BioScience
PE10611 |
50 mL | Research Grade |
£151.55
50mL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17987731
|
PL BioScience
PE10621 |
50 mL | GMP Grade |
£186.19
50mL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17967731
|
PL BioScience
PE10811 |
100 mL | Research Grade |
£259.80
100mL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17957741
|
PL BioScience
PE10821 |
100 mL | GMP Grade |
£333.41
100mL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17987721
|
PL BioScience
PE11011 |
500 mL | Research Grade |
£1212.40
500mL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17977721
|
PL BioScience
PE11021 |
500 mL | GMP Grade |
£1558.80
500mL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Xeno-Free: The xeno-free cell culture supplement is obtained from transfusion-approved platelets obtained at licensed blood centres.
- EU Origin: ELAREM™ Perform Research Grade is manufactured from platelet units obtained from healthy blood donors at licensed blood centres in the EU.
- Improved Cell Performance: Human Platelet Lysate contains abundant growth factors and cytokines derived from human platelets. Those nutrients have been shown to stimulate cellular proliferation and maintain phenotype and differentiation potential of various cells.
- Reproducible Results: Each batch of ELAREM™ Perform is produced from large pools of transfusion-approved platelet units to ensure batch-to-batch consistency and enable reproducible conditions. Grade & GMP GradeB
- From Research to Clinical Development: ELAREM™ Perform meets the requirements of academic and industrial cell expansion and assures an easy transition of research results into clinical applications.
- The product contains coagulation factors. To inhibit coagulation of the complete cell culture medium, it is recommended to add an anticoagulant, e.g. heparin, to the cell culture medium.
- Intended Use: ELAREM™ Perform is for in vitro and manufacturing use only. The product is not intended for direct therapeutic use. For safety precautions, please see appropriate Safety Data Sheet (SDS).
Specifications
Adherent Cell Culture, Suspension Cell Culture | |
Cell Culture Supplement | |
Sterile-filtered | |
Dry Ice |
Liquid | |
Sterile | |
Liquid, Human Platelet Lysate |