Learn More
PL BioScience ELAREM™ Ultimate-Fdi, Human Platelet Lysate, Fibrinogen-depleted, Viral inactivated
ELAREM™ Ultimate-FDi is a GMP, virus-inactivated and fibrinogen-depleted human Platelet Lysate of US origin. Ultimate-FDi is gamma-irradiated in order to comply with the highest safety guidelines for clinical applications.
Brand: PL BioScience UL40622
292.06 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.05000kg
Description
- Virus Inactivated for Highest Safety: The final product is virus-inactivated in order to comply with the guidelines for clinical applications. Virus panel testing is performed according to ICH and EMA guidelines (ICH Q5A (R1) and CPMP/BWP/268/95).
- GMP: The cell culture supplement is tested and released in confirmity with the relevant cGMP guidelines.
- Highly Efficient Cell Performance: Each batch of ELAREM™ Ultimate-FDi is produced from large pools of platelet units to ensure batch-to-batch consistency and enable reproducible conditions.
- No Heparin Addition Required: Due to the fibrinogen-depletion process, ELAREM™ Ultimate-FDi does not require anticoagulant addition. Traces of a xeno-free anticoagulant may be present in the human Platelet Lysate.
- Suited to Cellular Therapeutics: Due to the combination of highest safety standards and efficient cell perfomance, ELAREM™ Ultimate-FDi is suited to clinical trial and therapeutic cell manufacturing needs.
- US Origin: ELAREM™ Ultimate-FDi is a fibrinogen-depleted Human Platelet Lysate of US origin supporting in vitro expansion of various primary cells and cell lines.
- Intended Use: ELAREM™ Ultimate-FDi is for in vitro and manufacturing use only. The product is not intended for direct therapeutic use. For safety precautions, please see appropriate Safety Data Sheet (SDS).
Specifications
Yellow | |
Liquid | |
Sterile | |
Liquid, Human Platelet Lysate, Fibrinogen-depleted, Viral-inactivated | |
50 mL |
Adherent Cell Culture, Suspension Cell Culture | |
Cell Culture Supplement | |
Sterile-filtered | |
GMP Grade | |
Dry Ice |