missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PL BioScience ELAREM™ Ultimate-Fdi, Human Platelet Lysate, Fibrinogen-depleted, Viral inactivated
ELAREM™ Ultimate-FDi is a GMP, virus-inactivated and fibrinogen-depleted human Platelet Lysate of US origin. Ultimate-FDi is gamma-irradiated in order to comply with the highest safety guidelines for clinical applications.
£307.43 - £2559.03
Specifications
Color | Yellow |
---|---|
Culture Type | Adherent Cell Culture, Suspension Cell Culture |
Form | Liquid |
Product Type | Cell Culture Supplement |
Sterility | Sterile |
Product Code | Brand | Quantity | Purity or Quality Grade | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Purity or Quality Grade | Price | Quantity & Availability | ||||
17957731
|
PL BioScience
UL40622 |
50 mL | GMP Grade |
£307.43
50mL |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17987741
|
PL BioScience
UL40822 |
100 mL | GMP Grade |
£558.57
100mL |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17947731
|
PL BioScience
UL41022 |
500 mL | GMP Grade |
£2559.03
500mL |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Virus Inactivated for Highest Safety: The final product is virus-inactivated in order to comply with the guidelines for clinical applications. Virus panel testing is performed according to ICH and EMA guidelines (ICH Q5A (R1) and CPMP/BWP/268/95).
- GMP: The cell culture supplement is tested and released in confirmity with the relevant cGMP guidelines.
- Highly Efficient Cell Performance: Each batch of ELAREM™ Ultimate-FDi is produced from large pools of platelet units to ensure batch-to-batch consistency and enable reproducible conditions.
- No Heparin Addition Required: Due to the fibrinogen-depletion process, ELAREM™ Ultimate-FDi does not require anticoagulant addition. Traces of a xeno-free anticoagulant may be present in the human Platelet Lysate.
- Suited to Cellular Therapeutics: Due to the combination of highest safety standards and efficient cell perfomance, ELAREM™ Ultimate-FDi is suited to clinical trial and therapeutic cell manufacturing needs.
- US Origin: ELAREM™ Ultimate-FDi is a fibrinogen-depleted Human Platelet Lysate of US origin supporting in vitro expansion of various primary cells and cell lines.
- Intended Use: ELAREM™ Ultimate-FDi is for in vitro and manufacturing use only. The product is not intended for direct therapeutic use. For safety precautions, please see appropriate Safety Data Sheet (SDS).
Specifications
Yellow | |
Liquid | |
Sterile | |
Liquid, Human Platelet Lysate, Fibrinogen-depleted, Viral-inactivated | |
Dry Ice |
Adherent Cell Culture, Suspension Cell Culture | |
Cell Culture Supplement | |
Sterile-filtered | |
GMP Grade |