missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Solarus™ Electronic Burette

Digital, solar-powered display for easy reading of titration results. High accuracy with double seal PTFE compound piston.

Brand:  Hirschmann™ 9392050

 View more versions of this product

Product Code. 10093991

  • £1145.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For sensitive manual titration
  • Media conveying parts can be steam sterilized at 121°C
  • Free rotation
  • Media recirculating system for simple and rapid venting without reagent loss
  • Solar panels being the power supply for the display
  • Use within a temperature range of 4°C to 50°C
  • Individual quality certificate with conformity certificate
Specifications

Specifications

Autoclavable
50 mL
±0.20 mL
Electronic Buret
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hirschmann™ Solarus™ Electronic Burette > Capacity: 50 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.