missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Electronic Vial Crimpers and Decrimpers

Attach or remove crimp vial seals with high sample security and a simple, one-handed push of a button by using these Electronic Vial Crimpers and Decrimpers.

Brand:  Thermo Scientific™ 60180-ECRH13KIT

1795.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 16346218

  • £2585.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Tubes with 13 mm Crimp Caps
13 mm Electronic High Power Crimper Kit
Programmable or Manual Hand-Held Operation
13 mm
Electronic - Manual/Programmable
Electronic High Power Tool, 12 volt DC power supply with power cord, 13 mm Crimp Head, 13 mm Decrimp Head
13 mm aluminum, bi-metal, and steel caps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Electronic Vial Crimpers and Decrimpers >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.